Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145666.1 | NP_001139138.1 | 23 | 3234 | ||
NM_001145667.1 | NP_001139139.1 | 24 | 3267 | |||
NM_012201.5 | NP_036333.2 | 24 | 3267 | |||
NR_027264.1 | 25 | 3376 | ||||
NR_027265.1 | 19 | 2264 | ||||
GenBank mRNA | AK025457.1 | - | 21 | 2660 | ||
AK026972.1 | - | 15 | 1626 | |||
AK027032.1 | - | 25 | 3337 | |||
AK172806.1 | - | 7 | 728 | |||
AK304156.1 | - | 23 | 3204 | |||
BC031836.1 | - | 19 | 2218 | |||
BC060822.1 | - | 24 | 3238 | 5296427 | ||
U28811.1 | - | 24 | 3237 | |||
U64791.1 | - | 24 | 3220 |