Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005270.4 | NP_005261.2 | 12 | 2221 | ||
XM_006712422.2 | XP_006712485.1 | 12 | 2170 | |||
XM_011510969.2 | XP_011509271.2 | 12 | 2853 | |||
XM_011510970.2 | XP_011509272.1 | 12 | 3864 | |||
XM_011510971.2 | XP_011509273.1 | 12 | 3114 | |||
XM_011510972.2 | XP_011509274.2 | 12 | 2251 | |||
XM_011510973.2 | XP_011509275.1 | 11 | 2002 | |||
XM_011510974.2 | XP_011509276.1 | 11 | 2010 | |||
XM_017003818.1 | XP_016859307.1 | 12 | 2802 | |||
GenBank mRNA | AB007295.1 | - | 9 | 1778 | ||
AB007296.1 | - | 9 | 1727 | |||
AB007297.1 | - | 9 | 1778 | |||
AB007298.1 | - | 9 | 1727 | |||
AB209354.1 | - | 4 | 798 | |||
D14827.1 | - | 9 | 1727 | |||
D14828.1 | - | 8 | 1623 | |||
DQ086814.1 | - | 12 | 2191 |