Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005270.4 | NP_005261.2 | 4 | 561 | ||
XM_006712422.2 | XP_006712485.1 | 4 | 561 | |||
XM_011510969.2 | XP_011509271.2 | 4 | 1193 | |||
XM_011510970.2 | XP_011509272.1 | 4 | 2204 | |||
XM_011510971.2 | XP_011509273.1 | 4 | 1454 | |||
XM_011510972.2 | XP_011509274.2 | 4 | 591 | |||
XM_011510973.2 | XP_011509275.1 | 3 | 342 | |||
XM_011510974.2 | XP_011509276.1 | 3 | 401 | |||
XM_017003818.1 | XP_016859307.1 | 4 | 1193 | |||
GenBank mRNA | AY493737.1 | - | 4 | 507 | ||
BC111410.1 | - | 4 | 556 | |||
DQ004396.1 | - | 3 | 401 | |||
DQ086814.1 | - | 4 | 531 |