Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000168.5 | NP_000159.3 | 14 | 2295 | ||
XM_011515274.2 | XP_011513576.1 | 13 | 4919 | |||
XM_017011997.1 | XP_016867486.1 | 13 | 4952 | |||
GenBank mRNA | AK299299.1 | - | 13 | 2181 | ||
AK308429.1 | - | 15 | 2359 | |||
BC113616.1 | - | 13 | 2252 | 8322668 | ||
BC117168.1 | - | 13 | 2252 | 40125719 | ||
M57609.1 | - | 13 | 2258 |