Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203246.1 | NP_001190175.1 | 1 | 115 | ||
NM_022913.3 | NP_075064.1 | 1 | 115 | |||
XM_006714673.3 | XP_006714736.1 | 1 | 155 | |||
XM_006714674.3 | XP_006714737.1 | 1 | 155 | |||
XM_017009757.1 | XP_016865246.1 | 1 | 155 | |||
XM_017009758.1 | XP_016865247.1 | 1 | 155 | |||
GenBank mRNA | AK311625.1 | - | 1 | 115 | ||
AL161991.1 | - | 1 | 14 | |||
DB089516.1 | - | 1 | 116 | |||
DB271841.1 | - | 1 | 70 |