Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000168.5 | NP_000159.3 | 15 | 3573 | ||
XM_011515274.2 | XP_011513576.1 | 14 | 6197 | |||
XM_017011997.1 | XP_016867486.1 | 14 | 6230 | |||
GenBank mRNA | AK299299.1 | - | 14 | 3459 | ||
BC113616.1 | - | 14 | 3530 | 8322668 | ||
BC117168.1 | - | 14 | 3530 | 40125719 | ||
M57609.1 | - | 14 | 3536 |