Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001118885.1 | NP_001112357.1 | 2, 3 | 408 | ||
NM_001118886.1 | NP_001112358.1 | 1, 2 | 836 | |||
NM_002063.3 | NP_002054.1 | 1, 2 | 836 | |||
XM_017029427.1 | XP_016884916.1 | 2, 3 | 380 | |||
XM_017029428.1 | XP_016884917.1 | 1, 2 | 186 | |||
GenBank mRNA | AK289561.1 | - | 1, 2 | 363 | ||
AK312591.1 | - | 2, 3 | 408 | |||
AY437083.1 | - | 1, 2 | 76 | |||
AY437084.1 | - | 1, 2 | 76 | |||
BC032864.2 | - | 1, 2 | 606 | 5263356 | ||
CR450343.1 | - | 1, 2 | 76 | |||
X52008.1 | - | 1, 2 | 463 |