Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH10B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099697.1 | NP_001093167.1 | 20 | 2595 | ||
XM_005249833.3 | XP_005249890.1 | 14 | 1802 | |||
XM_006715766.2 | XP_006715829.1 | 21 | 2747 | |||
XM_006715768.2 | XP_006715831.1 | 14 | 1727 | |||
XM_011515491.1 | XP_011513793.1 | 20 | 2639 | |||
XM_011515493.1 | XP_011513795.1 | 20 | 2572 | |||
GenBank mRNA | AK294481.1 | - | 11 | 1460 | ||
AK314983.1 | - | 19 | 2715 | |||
BC044242.1 | - | 15 | 1940 |
Gene Symbol: RSPH10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173565.3 | NP_775836.3 | 20 | 2595 | ||
XM_005249659.3 | XP_005249716.1 | 20 | 3097 | |||
XM_011515203.1 | XP_011513505.1 | 21 | 2692 | |||
XM_011515204.2 | XP_011513506.1 | 20 | 2880 | |||
XM_011515207.1 | XP_011513509.1 | 20 | 2572 | |||
XM_011515208.2 | XP_011513510.1 | 18 | 2917 | |||
XM_011515209.2 | XP_011513511.1 | 18 | 2902 | |||
XM_011515210.1 | XP_011513512.1 | 14 | 1854 | |||
XM_011515211.1 | XP_011513513.1 | 14 | 1727 | |||
GenBank mRNA | BC034495.1 | - | 20 | 2598 | 4825868 | |
BC157864.1 | - | 18 | 2345 | 9057083 | ||
BC157871.1 | - | 18 | 2345 | 9057090 |