Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256310.1 | NP_001243239.1 | 6 | 1177 | ||
NM_014905.4 | NP_055720.3 | 6 | 1177 | |||
XM_005246467.3 | XP_005246524.1 | 6 | 1177 | |||
XM_006712435.1 | XP_006712498.1 | 6 | 1177 | |||
XM_017003829.1 | XP_016859318.1 | 6 | 1177 | |||
GenBank mRNA | AB020645.1 | - | 6 | 1163 | ||
AF097492.1 | - | 6 | 811 | |||
AF158555.1 | - | 7 | 1157 | |||
AF223943.1 | - | 6 | 932 | |||
AF279697.1 | - | 3 | 262 | |||
AF327434.1 | - | 6 | 1143 | |||
AK001220.1 | - | 3 | 249 | |||
AK289747.1 | - | 7 | 1153 | |||
BC038507.1 | - | 7 | 1153 | 5263220 |