Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014905.4 | NP_055720.3 | 17, 18 | 2120 | ||
GenBank mRNA | AB020645.1 | - | 17, 18 | 2106 | ||
AF097493.1 | - | 10, 11 | 848 | |||
AF223943.1 | - | 17, 18 | 1875 | |||
AF327434.1 | - | 17, 18 | 2086 | |||
AK092974.1 | - | 3, 4 | 1631 | |||
AK289747.1 | - | 18, 19 | 2096 | |||
AK294927.1 | - | 5, 6 | 738 | |||
AK298266.1 | - | 6, 7 | 472 | |||
BC038507.1 | - | 18, 19 | 2096 | 5263220 |