Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080396.2 | NP_001073865.1 | 2 | 1485 | ||
XM_011521109.2 | XP_011519411.1 | 2 | 1628 | |||
GenBank mRNA | AK295445.1 | - | 2 | 1191 | ||
BC146878.1 | - | 2 | 1086 | 9056806 | ||
BC157830.1 | - | 2 | 1086 | 9057049 | ||
BC157855.1 | - | 2 | 1086 | 9057074 | ||
BC157862.1 | - | 2 | 1086 | 9057081 | ||
L10374.1 | - | 2 | 1120 |