Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 19 | 3087 | ||
GenBank mRNA | AF100742.1 | - | 9 | 1082 | ||
AJ314790.1 | - | 19 | 3043 | |||
AK025481.1 | - | 7 | 780 | |||
AK055938.1 | - | 2 | 835 | |||
AK123306.1 | - | 3 | 1483 | |||
AK295297.1 | - | 6 | 633 | |||
AK307808.1 | - | 5 | 425 | |||
AL137258.1 | - | 2 | 102 | |||
BC051893.1 | - | 12 | 1678 | |||
BC062986.1 | - | 12 | 1603 | |||
BC137084.1 | - | 19 | 3051 | 9021081 | ||
BC137085.1 | - | 19 | 3051 | 9021082 |