Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002378.3 | NP_002369.2 | 9 | 1143 | ||
NM_139354.2 | NP_647611.1 | 8 | 979 | |||
NM_139355.2 | NP_647612.1 | 9 | 1202 | |||
XM_011528018.1 | XP_011526320.1 | 9 | 1357 | |||
GenBank mRNA | AB593100.1 | - | 9 | 1104 | ||
AK055395.1 | - | 9 | 1143 | |||
AK225834.1 | - | 8 | 802 | |||
AK315646.1 | - | 9 | 1095 | |||
AL137754.1 | - | 8 | 852 | |||
BC000114.1 | - | 9 | 1201 | 3350365 | ||
BC003109.1 | - | 9 | 1201 | 3357648 | ||
L18974.1 | - | 9 | 1064 | |||
S71669.1 | - | 8 | 800 | |||
X77278.1 | - | 9 | 1009 |