Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004124.2 | NP_004115.1 | 2 | 164 | ||
GenBank mRNA | AB001106.1 | - | 2 | 162 | ||
AB209240.1 | - | 2 | 96 | |||
AK311753.1 | - | 2 | 98 | |||
BC005359.1 | - | 2 | 118 | 3681628 | ||
BI547506.1 | - | 2 | 174 | |||
BT006784.1 | - | 1 | 65 | |||
BX647679.1 | - | 2 | 164 | |||
CR542113.1 | - | 1 | 65 | |||
M86492.1 | - | 2 | 105 |