Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006877.3 | NP_006868.3 | 3 | 343 | ||
XM_011514508.2 | XP_011512810.1 | 3 | 331 | |||
GenBank mRNA | BC008281.1 | - | 3 | 343 | 3635871 | |
M24470.1 | - | 3 | 300 |