Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002378.3 | NP_002369.2 | 9, 10 | 1176 | ||
NM_139354.2 | NP_647611.1 | 8, 9 | 1012 | |||
NM_139355.2 | NP_647612.1 | 9, 10 | 1235 | |||
XM_011528018.1 | XP_011526320.1 | 9, 10 | 1390 | |||
GenBank mRNA | AB593100.1 | - | 9, 10 | 1137 | ||
AK055395.1 | - | 9, 10 | 1176 | |||
AK225834.1 | - | 8, 9 | 835 | |||
AK315646.1 | - | 9, 10 | 1128 | |||
AL137754.1 | - | 8, 9 | 885 | |||
BC000114.1 | - | 9, 10 | 1234 | 3350365 | ||
BC003109.1 | - | 9, 10 | 1234 | 3357648 | ||
L18974.1 | - | 9, 10 | 1097 | |||
S71669.1 | - | 8, 9 | 833 | |||
X77278.1 | - | 9, 10 | 1042 |