Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029388.2 | 12, 13 | 1381 | |||
GenBank mRNA | AK293897.1 | - | 8, 9 | 909 | ||
AK311609.1 | - | 12, 13 | 1353 |