Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018089.2 | NP_001018099.1 | 8, 9 | 1038 | ||
NM_001276385.1 | NP_001263314.1 | 8, 9 | 1198 | |||
NM_024611.5 | NP_078887.2 | 8, 9 | 1198 | |||
XM_005254661.1 | XP_005254718.1 | 9, 10 | 1197 | |||
XM_005254662.1 | XP_005254719.1 | 9, 10 | 1060 | |||
XM_011522009.1 | XP_011520311.1 | 8, 9 | 1072 | |||
XM_011522010.1 | XP_011520312.1 | 8, 9 | 941 | |||
XM_011522011.2 | XP_011520313.1 | 7, 8 | 855 | |||
XM_017022569.1 | XP_016878058.1 | 9, 10 | 1234 | |||
GenBank mRNA | AF502591.1 | - | 8, 9 | 1141 | ||
AK055752.1 | - | 8, 9 | 1141 | |||
AK095389.1 | - | 8, 9 | 1022 | |||
AK127892.1 | - | 1, 2 | 23 | |||
AL832046.1 | - | 8, 9 | 1165 | |||
AY358546.1 | - | 8, 9 | 1159 | |||
BC020918.1 | - | 8, 9 | 1402 | |||
BC140894.1 | - | 8, 9 | 1117 | 9021762 | ||
BC151844.1 | - | 8, 9 | 1112 | |||
BX647873.1 | - | 8, 9 | 1024 |