Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025222.3 | NP_079498.2 | 5 | 756 | ||
XM_011534136.2 | XP_011532438.1 | 4 | 352 | |||
GenBank mRNA | AK098435.1 | - | 3 | 202 | ||
AK123860.1 | - | 3 | 1335 | |||
AK291380.1 | - | 5 | 512 | |||
AY358264.1 | - | 5 | 491 | |||
BX648628.1 | - | 5 | 498 |