Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256414.1 | NP_001243343.1 | 5 | 756 | ||
NM_002069.5 | NP_002060.4 | 5 | 893 | |||
GenBank mRNA | AF055013.1 | - | 5 | 568 | ||
AF493905.1 | - | 5 | 556 | |||
AK292953.1 | - | 5 | 867 | |||
AK304442.1 | - | 5 | 756 | |||
AL049933.1 | - | 5 | 871 | |||
BC026326.1 | - | 5 | 892 | 4791696 | ||
BT019775.1 | - | 5 | 556 | |||
M17219.1 | - | 5 | 541 |