Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 14 | 2395 | ||
GenBank mRNA | AB051501.2 | - | 14 | 2302 | ||
AF333769.2 | - | 14 | 2263 | |||
AK056833.1 | - | 14 | 2392 | |||
AK294281.1 | - | 12 | 1905 | |||
BC132737.1 | - | 14 | 2297 | 40146759 | ||
BC144130.1 | - | 12 | 2018 |
Gene Symbol: CNTNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201380.2 | NP_001188309.2 | 14 | 2566 |
Gene Symbol: CNTNAP3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111893.2 | 14 | 2255 |
Gene Symbol: LOC101930090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015388.1 | XP_016870877.1 | 5 | 740 |