Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020988.2 | NP_066268.1 | 3 | 1093 | ||
NM_138736.2 | NP_620073.2 | 3 | 1093 | |||
XM_011523003.2 | XP_011521305.1 | 3 | 451 | |||
GenBank mRNA | AF493894.1 | - | 3 | 196 | ||
AF493895.1 | - | 3 | 196 | |||
AK056008.1 | - | 3 | 1042 | |||
BC030027.2 | - | 3 | 129 | 4791738 | ||
CR936770.1 | - | 3 | 456 | |||
KR081949.1 | - | 3 | 196 | |||
M19182.1 | - | 1 | 36 |