Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037808.2 | NP_001032897.1 | 5 | 637 | ||
NM_001199142.1 | NP_001186071.1 | 5 | 757 | |||
NM_001267574.2 | NP_001254503.1 | 4 | 726 | |||
NM_001286478.1 | NP_001273407.1 | 5 | 558 | |||
NM_003752.4 | NP_003743.1 | 5 | 588 | |||
XM_017023814.1 | XP_016879303.1 | 5 | 543 | |||
GenBank mRNA | AK000739.1 | - | 4 | 618 | ||
AK055366.1 | - | 5 | 457 | |||
AK292155.1 | - | 5 | 757 | |||
AK301183.1 | - | 496 | ||||
AK303790.1 | - | 5 | 501 | |||
AK304500.1 | - | 4 | 267 | |||
AY762099.1 | - | 5 | 500 | |||
BC001571.1 | - | 5 | 522 | 3452953 | ||
BC071705.1 | - | 5 | 516 | 5552014 | ||
BC107692.1 | - | 5 | 548 | |||
BC157842.1 | - | 5 | 646 | 9057061 | ||
BC157849.1 | - | 4 | 486 | 9057068 | ||
BE780094.1 | - | 5 | 565 |
Gene Symbol: EIF3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099661.2 | NP_001093131.1 | 5 | 589 | ||
NM_001317856.1 | NP_001304785.1 | 5 | 634 | |||
NM_001317857.1 | NP_001304786.1 | 5 | 585 | |||
XM_017023620.1 | XP_016879109.1 | 5 | 541 | |||
GenBank mRNA | BQ686089.1 | - | 5 | 554 | ||
DC410988.1 | - | 5 | 585 |