Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099661.2 | NP_001093131.1 | 17 | 2431 | ||
NM_001317856.1 | NP_001304785.1 | 17 | 2476 | |||
NM_001317857.1 | NP_001304786.1 | 17 | 2427 | |||
XM_017023620.1 | XP_016879109.1 | 17 | 2383 | |||
XM_017023621.1 | XP_016879110.1 | 10 | 1558 |
Gene Symbol: EIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037808.2 | NP_001032897.1 | 17 | 2476 | ||
NM_001199142.1 | NP_001186071.1 | 17 | 2596 | |||
NM_001267574.2 | NP_001254503.1 | 16 | 2565 | |||
NM_001286478.1 | NP_001273407.1 | 17 | 2397 | |||
NM_003752.4 | NP_003743.1 | 17 | 2427 | |||
XM_017023814.1 | XP_016879303.1 | 17 | 2382 | |||
GenBank mRNA | AK000739.1 | - | 16 | 2457 | ||
AK055366.1 | - | 17 | 2296 | |||
AK130531.1 | - | 10 | 1624 | |||
AK292155.1 | - | 17 | 2596 | |||
AK293264.1 | - | 15 | 2005 | |||
AK299421.1 | - | 13 | 1836 | |||
AK301183.1 | - | 16 | 2335 | |||
AK301233.1 | - | 14 | 1923 | |||
AK303790.1 | - | 17 | 2340 | |||
AY762099.1 | - | 17 | 2339 | |||
BC000533.2 | - | 7 | 928 | |||
BC001571.1 | - | 17 | 2361 | 3452953 | ||
BC071705.1 | - | 17 | 2355 | 5552014 | ||
BC157842.1 | - | 17 | 2485 | 9057061 | ||
BC157849.1 | - | 16 | 2325 | 9057068 | ||
BT007335.1 | - | 3 | 531 | |||
BX648075.1 | - | 9 | 3429 |