Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317889.1 | NP_001304818.1 | 3, 4 | 172 | ||
NM_001317891.1 | NP_001304820.1 | 2, 3 | 106 | |||
NM_032026.3 | NP_114415.1 | 3, 4 | 172 | |||
NR_027427.1 | 3, 4 | 172 | ||||
XM_006716666.3 | XP_006716729.1 | 3, 4 | 192 | |||
XM_006716667.3 | XP_006716730.1 | 3, 4 | 161 | |||
XM_006716668.3 | XP_006716731.1 | 3, 4 | 174 | |||
XM_011517331.2 | XP_011515633.1 | 2, 3 | 88 | |||
XM_017013895.1 | XP_016869384.1 | 3, 4 | 182 | |||
XM_017013896.1 | XP_016869385.1 | 3, 4 | 161 | |||
XM_017013897.1 | XP_016869386.1 | 2, 3 | 82 | |||
XM_017013899.1 | XP_016869388.1 | 3, 4 | 168 | |||
GenBank mRNA | AF212250.1 | - | 3, 4 | 155 | ||
AK312204.1 | - | 3, 4 | 168 | |||
BC064964.1 | - | 3, 4 | 156 | 6141834 | ||
BG529611.1 | - | 2, 3 | 97 | |||
BX332564.2 | - | 3, 4 | 154 |