Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005272.3 | NP_005263.1 | 3 | 494 | ||
XM_011541264.2 | XP_011539566.1 | 4 | 585 | |||
GenBank mRNA | AF493909.1 | - | 3 | 281 | ||
BC000233.1 | - | 3 | 462 | 3350818 | ||
BT009844.1 | - | 3 | 281 |