Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004705.3 | NP_004696.2 | 5 | 2203 | ||
NR_130898.1 | 6 | 2298 | ||||
GenBank mRNA | AF081567.1 | - | 5 | 2074 | ||
AK001522.1 | - | 1 | 642 | |||
AK291843.1 | - | 5 | 2075 | |||
AK299594.1 | - | 3 | 1834 | |||
AL049970.1 | - | 1 | 1607 | |||
BC021992.1 | - | 1 | 493 | |||
BC110304.1 | - | 5 | 2180 | 5222582 | ||
BC117138.1 | - | 5 | 2132 | 40125689 | ||
BC117140.1 | - | 5 | 2132 | 40125691 | ||
BX641144.1 | - | 2 | 5674 |