Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282755.1 | NP_001269684.1 | 1 | 390 | ||
XM_005264459.2 | XP_005264516.1 | 1 | 390 | |||
GenBank mRNA | AK127245.1 | - | 1 | 390 | ||
DA239202.1 | - | 1 | 390 |