Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098721.1 | NP_001092191.1 | 4 | 886 | ||
NM_001098722.1 | NP_001092192.1 | 4 | 1012 | |||
NM_004485.3 | NP_004476.1 | 3 | 774 | |||
XM_006711761.2 | XP_006711824.1 | 3 | 657 | |||
XM_011544167.2 | XP_011542469.1 | 3 | 587 | |||
GenBank mRNA | BC022485.1 | - | 3 | 773 | 4799603 | |
U31382.1 | - | 3 | 656 |