Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005274.2 | NP_005265.1 | 3 | 457 | ||
GenBank mRNA | AF038955.1 | - | 2 | 140 | ||
AF493873.1 | - | 2 | 103 | |||
AK312111.1 | - | 2 | 216 | |||
BC003563.1 | - | 2 | 186 | 3502879 | ||
BM809260.1 | - | 2 | 196 | |||
BT006823.1 | - | 2 | 103 |