Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004126.3 | NP_004117.1 | 2 | 562 | ||
GenBank mRNA | AF493878.1 | - | 2 | 128 | ||
AK312037.1 | - | 2 | 212 | |||
BC009709.1 | - | 2 | 479 | 3879341 | ||
BG255463.1 | - | 572 | ||||
BT007141.1 | - | 2 | 128 | |||
U31384.1 | - | 2 | 235 |