Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031957.1 | NP_114163.1 | 1 | 710 | ||
GenBank mRNA | AJ406928.1 | - | 1 | 710 |