Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012009.1 | XP_016867498.1 | 3 | 266 | ||
GenBank mRNA | AF493869.1 | - | 2 | 131 | ||
BC029367.1 | - | 3 | 279 | 4604788 | ||
BG435040.1 | - | 3 | 279 | |||
BQ637754.1 | - | 3 | 178 | |||
CR456912.1 | - | 2 | 131 | |||
EL950965.1 | - | 3 | 206 | |||
S62027.1 | - | 2 | 139 |