Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005275.3 | NP_005266.2 | 2, 3 | 1623 | ||
XM_005249015.3 | XP_005249072.1 | 3, 4 | 1086 | |||
GenBank mRNA | AK026391.1 | - | 2, 3 | 285 | ||
AK298042.1 | - | 2, 3 | 462 | |||
AK301738.1 | - | 2, 3 | 213 | |||
AK302480.1 | - | 1, 2 | 351 | |||
AK316391.1 | - | 2, 3 | 289 | |||
BC013959.2 | - | 2, 3 | 272 | 2961417 | ||
BC018366.1 | - | 2, 3 | 1203 | 3924808 | ||
BT006648.1 | - | 2, 3 | 232 | |||
L25665.1 | - | 2, 3 | 186 |