Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000825.3 | NP_000816.4 | 3 | 2041 | ||
NM_001083111.1 | NP_001076580.1 | 4 | 1172 | |||
GenBank mRNA | M12578.1 | - | 4 | 363 | ||
X01059.1 | - | 3 | 1405 |