Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000406.2 | NP_000397.1 | 3 | 2512 | ||
NM_001012763.1 | NP_001012781.1 | 3 | 2384 | |||
GenBank mRNA | AY392011.1 | - | 3 | 761 | ||
BC113546.1 | - | 3 | 787 | 8322598 | ||
L03380.1 | - | 3 | 816 | |||
L07949.1 | - | 3 | 785 | |||
S60587.1 | - | 3 | 785 | |||
S77472.1 | - | 3 | 786 | |||
Z81148.1 | - | 3 | 633 |
Set Membership: |
G-protein coupled receptor |