Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VOPP1 SIRNA Assays ›
Gene Symbol:
VOPP1
Gene Name:
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases:
ECOP, GASP
Chromosome Location:
Chr.7: 55434406 - 55572520 on Build GRCh38
UniGene ID:
Hs.488307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s59351
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VOPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284282.1
NP_001271211.1
2
155
NM_001284283.1
NP_001271212.1
2
89
NM_001284284.1
NP_001271213.1
2
250
NM_001321242.1
NP_001308171.1
2
170
NM_001321243.1
NP_001308172.1
2
284
NM_001321244.1
NP_001308173.1
3
356
NM_001321246.1
NP_001308175.1
2
284
NM_001321247.1
NP_001308176.1
2
284
NM_001321248.1
NP_001308177.1
2
111
NM_001321249.1
NP_001308178.1
2
172
NM_001321250.1
NP_001308179.1
2
194
NM_001321251.1
NP_001308180.1
2
180
NM_030796.4
NP_110423.3
2
284
XM_011515539.1
XP_011513841.1
2
226
XM_011515540.2
XP_011513842.1
2
432
XM_011515541.2
XP_011513843.1
2
1125
XM_011515546.2
XP_011513848.1
2
954
GenBank mRNA
AB097004.1
-
2
143
AF395824.1
-
1
49
AK075391.1
-
2
218
AK090825.1
-
2
194
AK092992.1
-
2
228
AK225755.1
-
2
229
AK309288.1
-
2
155
AK314338.1
-
2
177
AK315882.1
-
2
82
AL136610.1
-
1
97
BC016650.1
-
1
176
4342536
BI597655.1
-
210
CN299440.1
-
2
290
CR533483.1
-
2
88
DA257111.1
-
2
177
DA530391.1
-
2
111
DA732543.1
-
3
123
DA814621.1
-
2
89
DB475432.1
-
2
289
DB496308.1
-
2
289
HY032103.1
-
2
173
HY057980.1
-
2
205
HY073578.1
-
2
254
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
signal transducer activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline