Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004486.4 | NP_004477.3 | 9, 10 | 672 | ||
XM_005251930.1 | XP_005251987.1 | 10, 11 | 771 | |||
XM_005251931.1 | XP_005251988.1 | 9, 10 | 690 | |||
XM_005251932.1 | XP_005251989.1 | 8, 9 | 602 | |||
GenBank mRNA | AF248953.1 | - | 9, 10 | 623 | ||
BC069268.1 | - | 9, 10 | 672 | 6340972 |