Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282301.1 | NP_001269230.1 | 1 | 1239 |
Gene Symbol: LOC100129307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310140.1 | NP_001297069.1 | 1 | 1358 |
Gene Symbol: LOC100131107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310142.1 | NP_001297071.1 | 1 | 1298 |
Gene Symbol: LOC100505502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519798.2 | XP_011518100.1 | 1 | 1209 | ||
GenBank mRNA | AK094743.1 | - | 2 | 1308 |