Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172557.1 | NP_001166028.1 | 5 | 967 | ||
NM_005895.3 | NP_005886.2 | 5 | 1305 | |||
XM_005266164.4 | XP_005266221.1 | 5 | 912 | |||
XM_005266167.4 | XP_005266224.1 | 2 | 337 | |||
XM_006719736.3 | XP_006719799.1 | 5 | 1032 | |||
XM_006719737.3 | XP_006719800.1 | 5 | 1042 | |||
XM_011534793.2 | XP_011533095.1 | 4 | 759 | |||
GenBank mRNA | AB027133.1 | - | 5 | 1027 | ||
AF485338.1 | - | 4 | 749 | |||
BC060826.1 | - | 5 | 991 | |||
BC142658.1 | - | 5 | 967 | 40148073 | ||
BC146675.1 | - | 5 | 967 | 40148852 | ||
D63997.1 | - | 5 | 1015 |