Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005895.3 | NP_005886.2 | 17 | 3850 | ||
XM_005266164.4 | XP_005266221.1 | 17 | 3457 | |||
XM_005266167.4 | XP_005266224.1 | 14 | 2882 | |||
XM_006719736.3 | XP_006719799.1 | 17 | 3577 | |||
XM_006719737.3 | XP_006719800.1 | 17 | 3587 | |||
XM_011534793.2 | XP_011533095.1 | 16 | 3304 | |||
XM_011534794.2 | XP_011533096.1 | 9 | 1455 | |||
GenBank mRNA | AB027133.1 | - | 17 | 3572 | ||
AF485338.1 | - | 16 | 3294 | |||
D63997.1 | - | 17 | 3560 | |||
L06148.1 | - | 7 | 984 |