Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172713.1 | NP_001166184.1 | 6 | 980 | ||
NM_002078.4 | NP_002069.2 | 5 | 914 | |||
XM_005265069.3 | XP_005265126.1 | 7 | 999 | |||
XM_005265070.3 | XP_005265127.1 | 7 | 999 | |||
XM_005265071.3 | XP_005265128.1 | 7 | 999 | |||
XM_005265072.3 | XP_005265129.1 | 6 | 951 | |||
XM_005265073.1 | XP_005265130.1 | 6 | 900 | |||
XM_005265074.1 | XP_005265131.1 | 6 | 900 | |||
XM_005265075.1 | XP_005265132.1 | 4 | 788 | |||
XM_011533598.2 | XP_011531900.1 | 1 | 42 | |||
XM_017006184.1 | XP_016861673.1 | 7 | 999 | |||
XM_017006185.1 | XP_016861674.1 | 7 | 1489 | |||
XM_017006186.1 | XP_016861675.1 | 7 | 668 | |||
XM_017006187.1 | XP_016861676.1 | 5 | 852 | |||
GenBank mRNA | AB209693.1 | - | 6 | 895 | ||
AK290550.1 | - | 5 | 836 | |||
AK300192.1 | - | 6 | 900 | |||
BC045543.1 | - | 8 | 1875 | |||
U41740.1 | - | 5 | 839 | |||
X82834.1 | - | 5 | 761 |