Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256486.1 | NP_001243415.1 | 8 | 938 | ||
NM_001256487.1 | NP_001243416.1 | 8 | 966 | |||
NM_001256488.1 | NP_001243417.1 | 7 | 843 | |||
NM_004487.4 | NP_004478.3 | 8 | 923 | |||
XM_005247371.4 | XP_005247428.1 | 8 | 1398 | |||
XM_005247372.4 | XP_005247429.1 | 8 | 974 | |||
XM_005247373.1 | XP_005247430.1 | 7 | 790 | |||
XM_006713587.1 | XP_006713650.1 | 8 | 913 | |||
XM_006713588.1 | XP_006713651.1 | 8 | 898 | |||
XM_006713589.1 | XP_006713652.1 | 8 | 895 | |||
XM_006713590.1 | XP_006713653.1 | 7 | 790 | |||
XM_006713591.1 | XP_006713654.1 | 6 | 673 | |||
XM_011512699.2 | XP_011511001.1 | 8 | 865 | |||
XM_017006189.1 | XP_016861678.1 | 8 | 1383 | |||
XM_017006190.1 | XP_016861679.1 | 8 | 1550 | |||
XM_017006191.1 | XP_016861680.1 | 7 | 1275 | |||
XM_017006192.1 | XP_016861681.1 | 8 | 959 | |||
XM_017006193.1 | XP_016861682.1 | 8 | 956 | |||
XM_017006194.1 | XP_016861683.1 | 7 | 851 | |||
XM_017006195.1 | XP_016861684.1 | 2 | 820 | |||
GenBank mRNA | AB371588.1 | - | 8 | 911 | ||
AB593126.1 | - | 7 | 817 | |||
AK296903.1 | - | 4 | 397 | |||
AK298986.1 | - | 5 | 489 | |||
AK300437.1 | - | 8 | 912 | |||
BC065301.1 | - | 7 | 714 | |||
D25542.1 | - | 8 | 921 | |||
X75304.1 | - | 8 | 911 |