Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014900.1 | XP_016870389.1 | 1 | 12 | ||
XM_017014914.1 | XP_016870403.1 | 1 | 72 | |||
XM_017014915.1 | XP_016870404.1 | 1 | 51 | |||
XM_017014916.1 | XP_016870405.1 | 1 | 58 |