Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136570.1 | 8 | 891 |
Gene Symbol: FAM230A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136560.1 | 4 | 877 |
Gene Symbol: LOC107987394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030163.1 | XP_016885652.1 | 8 | 1356 |