Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000408.4 | NP_000399.3 | 14, 15 | 2012 | ||
NM_001083112.2 | NP_001076581.2 | 14, 15 | 2238 | |||
XM_005246469.2 | XP_005246526.1 | 14, 15 | 1951 | |||
XM_011510977.2 | XP_011509279.1 | 14, 15 | 2006 | |||
XM_011510978.2 | XP_011509280.1 | 15, 16 | 2056 | |||
XM_011510980.2 | XP_011509282.1 | 9, 10 | 1237 | |||
XM_017003830.1 | XP_016859319.1 | 14, 15 | 2092 | |||
XM_017003831.1 | XP_016859320.1 | 13, 14 | 1901 | |||
GenBank mRNA | AB209399.1 | - | 14, 15 | 2007 | ||
AF311325.1 | - | 15, 16 | 2048 | |||
AK093198.1 | - | 15, 16 | 2047 | |||
AK290057.1 | - | 14, 15 | 2173 | |||
AK291065.1 | - | 14, 15 | 2251 | |||
AK292817.1 | - | 14, 15 | 2011 | |||
AK299645.1 | - | 10, 11 | 1353 | |||
U12424.1 | - | 14, 15 | 2001 | |||
U36310.1 | - | 14, 15 | 1981 |