Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000408.4 | NP_000399.3 | 15, 16 | 2087 | ||
NM_001083112.2 | NP_001076581.2 | 15, 16 | 2313 | |||
XM_005246469.2 | XP_005246526.1 | 15, 16 | 2026 | |||
XM_011510977.2 | XP_011509279.1 | 15, 16 | 2081 | |||
XM_011510978.2 | XP_011509280.1 | 16, 17 | 2131 | |||
XM_011510980.2 | XP_011509282.1 | 10, 11 | 1312 | |||
XM_017003830.1 | XP_016859319.1 | 15, 16 | 2167 | |||
XM_017003831.1 | XP_016859320.1 | 14, 15 | 1976 | |||
GenBank mRNA | AB209399.1 | - | 15, 16 | 2082 | ||
AF311325.1 | - | 16, 17 | 2123 | |||
AK093198.1 | - | 16, 17 | 2122 | |||
AK290057.1 | - | 15, 16 | 2248 | |||
AK291065.1 | - | 15, 16 | 2326 | |||
AK292817.1 | - | 15, 16 | 2086 | |||
AK299645.1 | - | 11, 12 | 1428 | |||
BC019874.1 | - | 10, 11 | 1241 | 4521072 | ||
U12424.1 | - | 15, 16 | 2076 | |||
U36310.1 | - | 15, 16 | 2056 |