Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018452.5 | NP_060922.2 | 3, 4 | 1098 | ||
GenBank mRNA | AF161405.1 | - | 3, 4 | 435 | ||
AF217510.1 | - | 3, 4 | 440 | |||
AK000891.1 | - | 3, 4 | 346 | |||
BC029130.1 | - | 3, 4 | 366 | 5182086 | ||
BC146893.1 | - | 3, 4 | 408 | 9056821 | ||
BC146895.1 | - | 3, 4 | 408 | 9056823 |