Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273032.3 | XP_005273089.1 | 1 | 244 | ||
XM_011544041.2 | XP_011542343.1 | 1 | 244 | |||
XM_011544043.2 | XP_011542345.1 | 1 | 244 | |||
XM_011544044.2 | XP_011542346.1 | 1 | 244 | |||
XM_011544045.2 | XP_011542347.1 | 1 | 244 | |||
XM_011544046.2 | XP_011542348.1 | 1 | 244 | |||
XM_011544047.2 | XP_011542349.1 | 1 | 244 | |||
XM_017000153.1 | XP_016855642.1 | 1 | 244 | |||
XM_017000155.1 | XP_016855644.1 | 1 | 244 | |||
XM_017000156.1 | XP_016855645.1 | 1 | 244 | |||
XM_017000158.1 | XP_016855647.1 | 1 | 244 | |||
XM_017000159.1 | XP_016855648.1 | 1 | 244 | |||
XM_017000161.1 | XP_016855650.1 | 1 | 244 | |||
XM_017000162.1 | XP_016855651.1 | 1 | 244 | |||
GenBank mRNA | AK056349.1 | - | 1 | 244 |
Gene Symbol: LOC107983951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544355.2 | XP_011542657.1 | 1 | 297 |