Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003334.3 | NP_003325.2 | 15 | 1902 | ||
NM_153280.2 | NP_695012.1 | 15 | 1830 | |||
XM_005272649.1 | XP_005272706.1 | 16 | 2280 | |||
XM_011543954.2 | XP_011542256.1 | 16 | 1922 | |||
XM_017029777.1 | XP_016885266.1 | 15 | 2997 | |||
XM_017029778.1 | XP_016885267.1 | 16 | 4171 | |||
XM_017029779.1 | XP_016885268.1 | 15 | 4679 | |||
XM_017029780.1 | XP_016885269.1 | 15 | 1787 | |||
XM_017029781.1 | XP_016885270.1 | 15 | 1835 | |||
GenBank mRNA | AK293161.1 | - | 4 | 494 | ||
AK296866.1 | - | 5 | 520 | |||
BC013041.2 | - | 15 | 1769 | 3543021 | ||
BC020261.1 | - | 12 | 1464 | |||
M58028.1 | - | 15 | 1808 | |||
X52897.1 | - | 8 | 974 | |||
X56976.1 | - | 15 | 1711 |